Learn More
TARGETMOL CHEMICALS INC CB2R PAM 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T370755MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CB2R PAM is an orally active cannabinoid type 2 receptor (CB2Rs) positive mutational modulator that enhances CP 55940 and 2-Arachidonylglycerol-stimulated [35S]GTP(gamma)S binding to CB2 receptors but has no effect in the absence of agonists. ...CB2R PAM shows anti-injury activity in a mouse model of neuropathic pain. purity: 98%
Catalog No. 50-258-0150