Learn More
BPS Bioscience Inc CBL-B TR-FRET ASSAY KIT
SDP
Supplier:  BPS Bioscience Inc 79575
Encompass
Human Casitas B-lineage lymphoma proto-oncogene b (CBL-B) is an E3 ubiquitin-protein ligase that functions as a negative regulator of T-cell activation. The CBL-B TR-FRET Assay Kit is designed to measure CBL-B auto-ubiquitination activity in a homogeneous 384 reaction format.
Catalog No. NC1889436