Learn More
Selleck Chemical LLC CBL0137 HCl 5mg 1197397-89-9 CBLC137 HCl, Curaxin 137 HCl
SDP
Supplier:  Selleck Chemical LLC S84835MG
Encompass_Preferred
CBL0137 (CBLC137, Curaxin 137) HCl activates p53 and inhibits NF-kB with EC50s of 0.37 ?M and 0.47 ?M in the cell-based p53 and NF-kB reporter assays, respectively. It also inhibits histone chaperone FACT (facilitates chromatin transcription complex). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8097