Learn More
Selleck Chemical LLC CBL0137 HCl
SDP
Supplier:  Selleck Chemical LLC S848325MG
Encompass_Preferred
CBL0137 (CBLC137 Curaxin 137) HCl activates p53 and inhibits NF- B with EC50s of 0 37 M and 0 47 M in the cell-based p53 and NF-kB reporter assays respectively It also inhibits histone chaperone FACT (facilitates chromatin transcription complex)
Catalog No. 50-000-01740