Learn More
ABclonal Technology CBR1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2356820UL
Encompass_Preferred
The protein encoded by this gene belongs to the short-chain dehydrogenases reductases (SDR) family which function as NADPH-dependent oxidoreductases having wide specificity for carbonyl compounds such as quinones prostaglandins and various xenobiotics Alternatively spliced transcript variants have been found for this gene
Catalog No. 50-000-06015