Learn More
STA PHARMACEUTICAL US LLC Cbz C2 amine amidite | 25 g | CAS 133339-58-9 | InChIKey BFXBTPRPHVBFNA-UHFFFAOYSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAM1071925GM
Cbz C2 amine amidite is a Amidite reagent (Subcategory: Amino Modifiers) sold by WuXi TIDES. Offered in 25 g. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: 133339-58-9
- MDL: No data
- InChIKey: BFXBTPRPHVBFNA-UHFFFAOYSA-N
- Molecular Weight: 395.439761998
- Molecular Formula: C19H30N3O4P
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29299090
- Country Of Origin: China
- IUPAC: benzyl (2-(((2-cyanoethoxy)(diisopropylamino)phosphaneyl)oxy)ethyl)carbamate
- SMILES: CC(C)N(C(C)C)P(OCCNC(OCC1=CC=CC=C1)=O)OCCC#N
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.