Learn More
Medchemexpress LLC Cbz-D-Glu(Bn)-Gly-Gly-Gly-Gly | 95.3% | C28H33N5O10 | 5 MG

Supplier: Medchemexpress LLC HY478125MG
Cbz-D-Glu(Bn)-Gly-Gly-Gly-Gly is a cleavable ADC linker, intended for research use only.
- Cleavable ADC linker
- Solid appearance
- White to off-white color
- Molecular weight: 599.59
- Store at -20°C, protected from light. In solvent, store at -80°C for 6 months, or -20°C for 1 month, protected from light.
- Shipping at room temperature in continental US; may vary elsewhere.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.