Learn More
Medchemexpress LLC Cbz-N-methyl-L-alanine | 21691-41-8 | MFCD00153386 | 99.4% | 237.25 g·mol⁻1 | C12H15NO4 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW022444100G
Encompass_Preferred
Z-N-Me-Ala-OH (Cbz-N-methyl-L-alanine) is a Cbz-protected N-methyl derivative of L-alanine supplied as a white to off-white solid. It serves as an amino acid building block for peptide synthesis and organic synthesis requiring N-methylated residues. Typical reported purity is ~99.4% and the molecular weight is 237.25 g·mol⁻1.
  • Cbz-protected N-methyl amino acid suitable for peptide coupling
  • High purity ideal for research and synthetic applications
  • White to off-white solid for easy handling and dosing
  • Stable when stored as powder at -20°C or 4°C per product guidance
  • Available in multiple pack sizes including 100 g for scale-up
Catalog No. 50-003-00120