Learn More
Medchemexpress LLC Cbz-NH-PEG4-C2-acid | 756526-00-8 | MFCD11041139 | 95.0% | 399.4 g/mol | C19H29NO8 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0197961G
Encompass_Preferred
Cbz-NH-PEG4-C2-acid is a Cbz-protected PEG4 linker bearing a two-carbon carboxylic acid terminus and a benzyloxycarbonyl-protected amine. It functions as a flexible, hydrophilic spacer and synthetic building block for assembling proteolysis targeting chimeras (PROTACs) and other small-molecule bioconjugates, enabling orthogonal coupling strategies in multi-step syntheses.
  • Cbz-protected amine for orthogonal coupling.
  • Four-unit polyethylene glycol spacer for flexibility and solubility.
  • Terminal two-carbon carboxylic acid for standard coupling chemistries.
  • Suitable for PROTAC synthesis and small-molecule conjugation.
  • Reported high purity to support reliable synthetic workflows.
Catalog No. 50-003-01851