Learn More
Medchemexpress LLC Cbz-NH-PEG4-C2-acid | 756526-00-8 | MFCD11041139 | >95.0% | 399.44 g·mol⁻¹ | C19H29NO8 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW019796100MG
Encompass_Preferred
Cbz-NH-PEG4-C2-acid is a Cbz-protected PEG-based linker containing a PEG4 spacer and a two-carbon carboxylic acid terminus, intended for use in PROTAC synthesis and related medicinal chemistry applications. The protected amine allows selective deprotection and downstream coupling to assemble bifunctional molecules.
  • Cbz-protected amine for selective deprotection.
  • PEG4 spacer provides linker flexibility and improved solubility.
  • C2 carboxylic acid terminus enables standard coupling chemistries.
  • Suitable for PROTAC and bioconjugation synthesis workflows.
  • Available in multiple pack sizes for research-scale use.
  • Characterized by molecular formula and molecular weight; consult SDS for purity details.
Catalog No. 50-003-01490