Learn More
Medchemexpress LLC Cbz-peg2-bromide | 2100283-00-7 | MFCD31916729 | 97.0% | 346.22 | C14H20BrNO4 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW190968250MG
Encompass_Preferred
Cbz-PEG2-bromide is a carbamate-protected PEG2-based bromide linker designed for the synthesis of proteolysis targeting chimeras (PROTACs) and other bifunctional molecules. It provides a short, flexible polyethylene glycol spacer with a reactive bromide terminus and a Cbz-protected amine, enabling orthogonal coupling strategies during multistep synthesis. Supplied at 97.0% purity in a 250 mg package.
  • Short, flexible PEG2 spacer for maintaining optimal linker length.
  • Reactive bromide terminus for efficient nucleophilic substitution reactions.
  • Cbz-protected amine enables orthogonal deprotection and coupling steps.
  • High purity (97.0%) suitable for research synthesis applications.
  • Stable under recommended storage conditions for long-term use.
  • Supplied in a 250 mg package convenient for small-scale synthesis.
Catalog No. 50-002-83180