Learn More
Medchemexpress LLC Cbz-TRIS tri-acid | 200133-16-0 | 471.46 | 10 G

Supplier: Medchemexpress LLC HY4431210G
Cbz-TRIS tri-acid is a small molecule compound that contains multiple functional groups, specifically a carbamate group, a tris group, and three carboxylic acid groups. This product is intended for research use only and is not sold to patients.
- Chemical structure: features a carbamate group, a tris group, and three carboxylic acid groups.
- Appearance: solid.
- Color: white to off-white.
- Shipping: ships at room temperature within the continental US, with potential variations elsewhere.
- Storage: can be stored at room temperature for 3 years; in solvent, it can be stored at -80°C for 2 years or -20°C for 1 year.
- Target: biochemical assay reagents.
- Pathway: others.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.