Learn More
Medchemexpress LLC Cbz-TRIS tri-acid | 200133-16-0 | 471.46 | 5 G
SDP
Supplier:  Medchemexpress LLC HY443125G
Encompass_Preferred
Cbz-TRIS tri-acid is a small molecule compound containing several functional groups, including a carbamate group, a tris group, and three carboxylic acid groups. This product is for research use only.
  • Room temperature shipping in continental US
  • Stable at room temperature for 3 years
  • Stable in solvent at -80°C for 2 years
  • Stable in solvent at -20°C for 1 year
Catalog No. 50-004-03610