Learn More
Medchemexpress LLC Cbz-Valaciclovir | 124832-31-1 | 98.1% | 458.47 | 250 MG

Supplier: Medchemexpress LLC 250MGHY126375
Cbz-Valaciclovir (Benzyloxycarbonyl valacyclovir) is an intermediate used to prepare Valaciclovir (HY-17425) and is intended for antiviral research. It is for research use only.
- Used for antiviral research
- Research use only, not sold to patients
- Appearance as a white to off-white solid
- Soluble in DMSO at 100 mg/mL
- Store at 4°C under nitrogen, away from moisture
- In solvent, store at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.