Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CC-3240 | 2922675-91-8 | 98.2% | 917.17 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY16306450MG

Encompass_Preferred

CC-3240 (compound 13) is a molecular glue degrader of CaMKK2 based on CC-8977, with an IC50 of 9 nM. It is intended for research use only.

  • Appearance: Solid
  • Color: White to off-white
  • Formula: C52H64N6O7S
  • Shipping conditions: Room temperature in continental US; may vary elsewhere
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 100 mg/mL (109.03 mM)

Catalog No. 50-003-92182


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.