Learn More
TARGETMOL CHEMICALS INC CC-401 HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T58335MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CC-401 Hydrochloride (CC401 HCl) is a second generation ATP-competitive anthrapyrazolone c-Jun N terminal kinase (JNK) inhibitor (Ki of 25 to 50 nM.)with potential antineoplastic activity. purity: 98%
Catalog No. 50-257-6657