Learn More
Medchemexpress LLC CC-671 | 1618658-88-0 | 99.1% | 512.56 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10870910MM/1ML
Encompass_Preferred
CC-671 is a dual TTK protein kinase/CDC2-like kinase (CLK2) inhibitor, demonstrating potent inhibitory activity against both enzymes. It exhibits cytotoxicity against human BT474 and CAL51 cells and shows significant tumor growth inhibition in vivo. This compound also selectively binds to kinases such as CLK2, CAMKK2, PIP4K22, and JNK.
  • Dual TTK protein kinase/CDC2-like kinase (CLK2) inhibitor
  • Exhibits cytotoxicity against human BT474 and CAL51 cells
  • Demonstrates significant tumor growth inhibition in vivo
  • Selectively binds to CLK2, CAMKK2, PIP4K22, and JNK
Catalog No. 50-003-46408