Learn More
Medchemexpress LLC HY-101488 50mg , CC-885 CAS:1010100-07-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10148850MG
Encompass_Preferred
HY-101488 50mg CC-885 CAS: 1010100-07-8 CC-885 is a cereblon (CRBN) modulator with potent anti-tumour activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0402