Learn More
Medchemexpress LLC HY-112570 5mg Medchemexpress, CC-90003 CAS:1621999-82-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1125705MG
Encompass_Preferred
Medchemexpress, HY-112570 5mg CC-90003 CAS:1621999-82-3 CC-90003 is an irreversible and selective inhibitor of ERK 1/2 with antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0820