Learn More
Medchemexpress LLC HY-139504 5mg , CC-99677 CAS:1887069-10-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1395045MG
Encompass_Preferred
HY-139504 5mg CC-99677 CAS: 1887069-10-4 CC-99677 is a potent, covalent, and irreversible inhibitor of the mitogen-activated protein (MAP) kinase-activated protein kinase-2 (MK2) pathway in both biochemical (IC50=156.3 nM) and cell based assays (EC50=89 nM). CC-99677 is extracted from patent WO2020236636, compound 1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9727