Learn More
Beckman Coulter Multisizer 4e
Supplier:  Beckman Coulter C72513
Encompass_Preferred
The Beckman Coulter Multisizer 4e is a benchtop particle counter using electrical sensing to size and count particles from 0.4 to 1,200 microns. It analyzes up to 32 samples at once with precision, featuring a touchscreen, 21 CFR Part 11 compliance, and detailed particle analysis. It's ideal for quality control in various industries.
Catalog No. NC2291478