Learn More
Medchemexpress LLC CC214-1 | 1021920-32-0 | C20H21N7O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15491010MM/1ML
Encompass_Preferred
CC214-1 is an efficacious mTOR inhibitor that induces autophagy with an IC50 of 0.002 μM. This compound serves as an in vitro tool for exploring mTOR kinase biology and can be used in glioblastoma studies.
  • Efficacious mTOR inhibitor
  • Induces autophagy
  • IC50 of 0.002 μM for mTOR
  • Inhibits mTORC2 and mTORC1 signaling in PC-3 cells
  • Can be used for glioblastoma studies
  • Synergizes with rapamycin to inhibit tumor cell proliferation
  • Efficiently inhibits T cell activation
Catalog No. 50-003-76204