Learn More
Medchemexpress LLC CC214-1 | 1021920-32-0 | C20H21N7O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY154910
Encompass_Preferred
CC214-1 is a potent mTOR inhibitor that induces autophagy, with an IC50 of 0.002 μM. This compound is useful as an in vitro tool for exploring mTOR kinase biology and for Glioblastoma studies.
  • Potent mTOR inhibitor (IC50 of 0.002 μM)
  • Induces autophagy
  • Useful for exploring mTOR kinase biology in vitro
  • Suitable for glioblastoma studies
  • Inhibits mTORC2 (IC50 of 0.018 μM in PC-3 cells) and mTORC1 (IC50 of 0.04 μM in PC-3 cells)
  • Synergizes with rapamycin to inhibit mTORC1 signaling and tumor cell proliferation
  • Mediates sensitivity to growth arrest in glioblastoma cells
  • Inhibits T cell activation and expression of activation markers
Catalog No. 50-003-75289