Learn More
Medchemexpress LLC CCC-0975 | 432018-03-6 | 99.5% | 483.89 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14915810MM/1ML
Encompass_Preferred
CCC-0975 is a hepatitis B virus (HBV) inhibitor with an EC50 of 10 μM. It specifically interferes with the conversion of relaxed circular DNA (rcDNA) to covalently closed circular DNA (cccDNA), leading to a synchronous reduction of both cccDNA and its precursor, deproteinized rcDNA (DP-rcDNA). This inhibition occurs without promoting their intracellular degradation, making CCC-0975 a promising compound for chronic hepatitis B research.
  • functions as a hepatitis B virus (HBV) inhibitor
  • interferes with rcDNA to cccDNA conversion
  • reduces cccDNA and deproteinized rcDNA levels
  • does not promote intracellular degradation
  • shows promise for chronic hepatitis B research
Catalog No. 50-003-61014