Learn More
Medchemexpress LLC CCC-0975 | 432018-03-6 | 99.5% | 483.89 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14915850MG
Encompass_Preferred
This compound is an inhibitor of the hepatitis B virus (HBV), demonstrating an EC50 of 10 μM. It works by interfering with the conversion process of relaxed circular DNA (rcDNA) into covalently closed circular DNA (cccDNA). This action leads to a synchronous reduction in both cccDNA and its precursor, deproteinized rcDNA (DP-rcDNA), without causing their degradation within the cell. This makes it a promising agent for the study of chronic hepatitis B.
Features and Benefits:
  • Specifically inhibits hepatitis B virus (HBV) with an EC50 of 10 μM.
  • Interferes with the conversion of rcDNA to cccDNA.
  • Reduces both cccDNA and DP-rcDNA levels.
  • Does not promote intracellular degradation of cccDNA or DP-rcDNA.
  • Potentially useful for research into chronic hepatitis B.
Catalog No. 50-003-60889