Learn More
Medchemexpress LLC Ccc R08 | 2919019-72-8 | 98.87% | 414.84 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14856010MM/1ML
Encompass_Preferred
Ccc_R08 is a non-cytotoxic and orally active cccDNA inhibitor that reduces cccDNA levels in the liver of HBV-infected mice. It can be used in the study of HBV virus (hepatitis B virus) infection.
  • Reduces cccDNA, protein-free RC-DNA, and double stranded linear DNA (DL-DNA) levels in HepDES19 cells.
  • Decreases serum levels of pgRNA in a dose-dependent manner.
  • Leads to the clearance of cccDNA from HBVcircle mouse livers.
Catalog No. 50-003-60570