Learn More
Medchemexpress LLC HY-100941 100mg , CCCP CAS:555-60-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100941100MG
Encompass_Preferred
HY-100941 100mg CCCP CAS: 555-60-2 CCCP is an oxidative phosphorylation (OXPHOS) uncoupler. CCCP induces activation of PINK1 leading to Parkin Ser65 phosphorylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9420