Learn More
Selleck Chemical LLC CCCP 25mg
SDP
Supplier:  Selleck Chemical LLC S649425MG
Encompass_Preferred
CCCP (Carbonyl cyanide m-chlorophenyl hydrazone, Carbonyl cyanide 3-chlorophenylhydrazone), anoxidative phosphorylation inhibitor, is a protonophore mitochondrial uncoupler that increases membrane permeability to protons, leading to a disruption in the mitochondrial membrane potential. Carbonyl cyanide 3-chlorophenylhydrazone (CCCP), the protonophore, can inhibitsSTING-mediated IFN-? production via disrupting mitochondrial membrane potential (MMP). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3181
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.