Learn More
ATCC CCD-8Lu; Lung Fibroblast; Human (Homo sapiens)
Supplier:  ATCC CCL201
Encompass
CCD-8Lu fibroblasts are cells derived from normal tissue from a patient who died of cerebral thrombosis. The cells senesce at about population doubling level (PDL) 40 to 42.
Catalog No. 50-238-1836