Learn More
Aviva Systems Biology Corp CCDC96 Antibody - N-terminal region (ARP55593_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55593_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC96. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CCDC96. Alternate names: FLJ90575, MGC138479ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61550