Learn More
Medchemexpress LLC Ccf0058981 | 2708934-53-4 | 98.0% | 442.90 g/mol | C24H19ClN6O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1323065MG
Encompass_Preferred
CCF0058981 is a small-molecule, noncovalent inhibitor of the SARS-CoV-2 3CL protease with potent enzymatic activity (IC50 = 68 nM) and reported antiviral efficacy. Molecular formula C24H19ClN6O, molecular weight 442.90 g/mol, CAS 2708934-53-4. Intended for laboratory research use only; follow recommended storage and handling instructions.
Catalog No. 50-002-62967