Learn More
TARGETMOL CHEMICALS INC CCF0058981 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T600955MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CCF0058981 (CCF981) is a 3-chlorophenyl analogue which is a noncovalent SARS-CoV-2 3CL pro (SC2) inhibitor with an IC 50 of 68 nM. CCF0058981 inhibits SC1 (SARS-CoV-1 3CL pro ) with an IC 50 of 19 nM. CCF0058981 has antiviral efficacy and has the potential for COVID-19 research [1]. purity: 98%
Catalog No. 50-257-9677