Learn More
TARGETMOL CHEMICALS INC CCG215022 2MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T34982MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. CCG215022 is a G protein-coupled receptor kinases (GRKs) inhibitor with IC50s of 0.15±0.07 uM for GRK2, 0.38±0.06 uM for GRK5, and 3.9±1 uM for GRK1. Purity 97.46%
Catalog No. 50-287-6331