Learn More
Medchemexpress LLC CCI-006 100mg | 292053-42-0 | 100 MG

Supplier: Medchemexpress LLC HY114410100MG
CCI-006 is a small-molecule inhibitor of mitochondrial metabolism that functions as a chemosensitizer in MLL-rearranged leukemia models. Supplied as a high-purity solid, it is intended for research use only.
- Selective inhibitor and chemosensitizer of MLL-rearranged leukemia cells.
- Molecular formula: C15H12N2O5S.
- Molecular weight: 332.33 g·mol-1.
- High purity (reported 99.87%).
- Physical form: solid powder.
- Recommended storage: powder at -20°C or 4°C; in solvent at -80°C.
- Intended for research use only; not for human consumption.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.