Learn More
Raybiotech Inc CCK EIA
Supplier:  Raybiotech Inc EIACCK1
Encompass_Preferred
RayBio Human/Mouse/Rat Cholecystokinin EIA Kit optimized for serum and cell culture medium. Competition-based ELISA on a 96-well strip plate.
Catalog No. 50-104-3263