Learn More
Cayman Chemical CCK27-33non-sulfated 1mg
SDP
Supplier:  Cayman Chemical 249521 MG
Encompass_Preferred
A C-terminal fragment of CCK; inhibits binding of [3H]naloxone in rat cerebellum membranes (IC50 = 4 µM); inhibits electrically-stimulated contraction of isolated guinea pig ileum (IC50 = 17 µM)
Catalog No. 50-293-5665