Learn More
ATCC ATCC L1210 (CCL-219) Mouse lymphocytic leukemia cell line
Supplier: ATCC L1210
L1210 (ATCC CCL-219) is a mouse lymphocytic leukemia cell line exhibiting lymphoblast morphology, isolated from the skin of an 8-month-old female mouse. It is commonly used for antitumor screening and immune system disorder research.
- Mouse lymphocytic leukemia cell line exhibiting lymphoblast morphology.
- Suitable for suspension culture at 37°C with 5% CO2.
- Grow in Dulbecco's Modified Eagle's Medium supplemented with 10% horse serum.
- Biosafety level 1; store frozen in vapor phase of liquid nitrogen.
- Cryopreserve using complete medium with 5% (v/v) DMSO.
- Seed at 5 x 10^4 viable cells/mL; refresh 20-30% medium every 2-4 days.
- Tested negative for ectromelia virus.