Learn More
ATCC K-562 are lymphoblast cells isolated from the bone marrow of a 53-year-old chronic myelogenous leukemia patient. The K-562 cell line is widely used in immune system disorder and immunology research.
Supplier:  ATCC K562
Encompass
K-562 are lymphoblast cells isolated from the bone marrow of a 53-year-old chronic myelogenous leukemia patient. The K-562 cell line is widely used in immune system disorder and immunology research.
Catalog No. NC3432035