Learn More
Medchemexpress LLC CCR2-RA-[R] | 512177-83-2 | 99.7% | 351.80 | 10 MG

Supplier: Medchemexpress LLC 10MGHY50081
CCR2-RA-[R] is an allosteric antagonist of the C-C chemokine receptor type 2 (CCR2), inhibiting it non-competitively with an IC50 of 103 nM. It functions by blocking activation-associated conformational changes and the formation of the G protein-binding interface.
- Binding pocket has balanced hydrophobic and polar features, favoring druggability
- Displaces [125I]CCL2 from CCR2 with pIC50 of 6.1
- pKD values for CCR2 and CCR5 are 8.8±0.1 and 7.0±0.1, respectively
- Soluble in DMSO (37.5 mg/mL)
- Stable as powder at -20°C for 3 years or 4°C for 2 years
- Stable in solvent at -80°C for 2 years or -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.