Learn More
Medchemexpress LLC CCR4-351 hydrochloride | 2174938-71-5 | 98.4% | C24H28Cl3N7O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY131349A5MG
Encompass_Preferred
CCR4-351 hydrochloride is an orally active, potent, and selective antagonist of the C-C chemokine receptor 4 (CCR4). The hydrochloride salt exhibits low-nanomolar cellular potency (≈22 nM in calcium flux assays, ≈50 nM in CTX assays) and has demonstrated antitumor activity in preclinical studies. It is supplied as a high-purity research compound intended for in vitro and exploratory in vivo pharmacology.
  • Orally active CCR4 antagonist with low-nanomolar potency.
  • Hydrochloride salt form for improved solubility and handling.
  • Demonstrated antitumor activity in preclinical models.
  • High purity suitable for research applications (≈98.4%).
  • Available in small-mass quantities for screening and pharmacology studies.
Catalog No. 50-002-62538