Learn More
Selleck Chemical LLC CCS-14775mg
SDP
Supplier:  Selleck Chemical LLC S96675MG
Encompass_Preferred
CCS1477(CBP-IN-1, CBP/p300-IN-4)is a potent and selective inhibitor of p300/CBP bromodomain. CCS1477 works by inhibiting the expression and function of the androgen receptor (AR), as well as inhibiting c-Myc. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3488