Learn More
TARGETMOL CHEMICALS INC CCT020312 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T149025MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CCT020312 is a selective activator of EIF2AK3 and PERK. CCT020312 durably inhibits cell proliferation and elicits the phosphorylation of EIF2A. purity: 98%
Catalog No. 50-258-1274