Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CCT031374 hydrobromide | 1219184-91-4 | MFCD06737234 | 98.1% | 434.33 g·mol⁻¹ | C23H20BrN3O | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1084415MG

Encompass_Preferred

CCT031374 hydrobromide is the hydrobromide salt of a small-molecule β-catenin/TCF signaling inhibitor supplied for research use only. It is provided as an off-white to light yellow solid with high chemical purity and is intended for pathway inhibition, cell signaling assays, and preclinical studies.

  • β-catenin/TCF signaling inhibitor
  • High purity (98.1%)
  • Molecular weight 434.33 g·mol⁻¹
  • Off-white to light yellow solid form
  • Available in milligram-scale packages for research use
  • Intended for in vitro and in vivo research applications

Catalog No. 50-002-63799


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.