Learn More
Selleck Chemical LLC CCT128930 5mg 885499-61-6
SDP
Supplier:  Selleck Chemical LLC S26355MG
Encompass_Preferred
CCT128930 is a potent, ATP-competitive and selective inhibitor of Akt2 with IC50 of 6 nM in a cell-free assay, 28-fold greater selectivity for Akt2 than the closely related PKA kinase. CCT128930 induces cell cycle arrest, DNA damage, and autophagy independent of Akt inhibition. High dose of CCT128930 triggers cell apoptosis in HepG2 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0534129