Learn More
Selleck Chemical LLC CCT137690 10mg 1095382-05-0
SDP
Supplier:  Selleck Chemical LLC S274410MG
Encompass_Preferred
CCT137690 is a highly selective inhibitor of Aurora A, Aurora B and Aurora C with IC50 of 15 nM, 25 nM and 19 nM. It has little effect on hERG ion-channel. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1332495