Learn More
TARGETMOL CHEMICALS INC CCT241161 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T96385MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CCT241161 is an orally active pan-RAF inhibitor with IC50 values of 3 6 10 15 and 30 nM for LCK CRAF SRC V600E-BRAF and BRAF respectively. It displays significant activity against BRAF and NRAS mutant melanomas as well as demonstrating anticancer cell proliferative effects [1]. purity: 98%
Catalog No. 50-257-8991