Learn More
Selleck Chemical LLC CCT241533 hydrochloride-1MG
SDP
Supplier:  Selleck Chemical LLC E16531MG
Encompass_Preferred
CCT241533 hydrochloride is an ATP competitive potent and selective inhibitor of CHK2 with an IC50 of 3 nM and a Ki of 1 16 nM and shows minimal cross-reactivity against a panel of kinases CCT241533 inhibits CHK2 activity in human tumor cell lines when subjected to DNA damage
Catalog No. 50-313-8548