Learn More
TARGETMOL CHEMICALS INC CCT245737 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T708010MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CCT245737 (SRA737) is an orally active and selective Chk1 inhibitor (IC50 1.3 nM); CCT245737 shows much less activity against Chk2 (IC50 2440 nM). purity: 99%
Catalog No. 50-257-6357