Learn More
Selleck Chemical LLC CCT251545 5mg 1661839-45-7
SDP
Supplier:  Selleck Chemical LLC S79815MG
Encompass_Preferred
CCT251545 is a potent, orally bioavailable inhibitor of WNT signaling with IC50 of 5 nM in 7dF3 cells. CCT251545 also act as a selective chemical probe for exploring the role of CDK8 and CDK19 in human disease. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1901473