Learn More
Medchemexpress LLC CCT369260 | 2647503-57-7 | >99.67% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12918810MG
Encompass_Preferred
CCT369260 is a chemical probe and an orally active B-cell lymphoma 6 (BCL6) inhibitor with anti-tumor activity. It exhibits an IC50 of 520 nM. In an in vivo study, CCT369260 (15 mg/kg, orally, single dose) significantly inhibited BCL6 in an OCI-Ly1 DLBCL xenograft model in female SCID mice, leading to decreased levels of BCL6 in the tumor observed up to 10 hours after administration.
  • Orally active B-cell lymphoma 6 (BCL6) inhibitor
  • Demonstrates anti-tumor activity
  • Exhibits an IC50 of 520 nM
  • Significant inhibition of BCL6 in OCI-Ly1 DLBCL xenograft model
Catalog No. 50-003-38818